Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-MMP-2 Clone: 42-5D11,

Catalog No. IM33100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
IM33100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IM33100UG Supplier MilliporeSigma™ Supplier No. IM33100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects MMP-2 Clone: 42-5D11 in Bovine, Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immuno Blotting

Matrix metalloproteinases (MMPs) are a family of enzymes that are responsible for the degradation of extracellular matrix components such as collagen, laminin, and proteoglycans. In addition to sequence homology, all MMPs share the following characteristics: the catalytic mechanism is dependent upon a zinc ion at the active center, they cleave one or more extracellular matrix components, they are secreted as zymogens which are activated by removal of an approximately 10kDa segment from the N-terminus, and they are inhibited by tissue inhibitor of metalloproteinases (TIMP). These enzymes are involved in normal physiological processes such as embryogenesis and tissue remodeling and may play an important role in arthritis, periodontitis, and metastasis. MMP-2 (Gelatinase A, 72kDa gelatinase/type IV collagenase, TBE-1) is secreted as a 72kDa zymogen which is proteolytically processed to the 66kDa active form. MMP-2, along with its most closely related member of the MMP family, MMP-9, show substrate specificity toward type IV and V collagens, gelatin, and elastin. Numerous studies have shown a correlation between collagenase expression and metastatic potential, and suggest that it may be a useful marker for the diagnosis or prognosis of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-2
Applications Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 42-5D11
Conjugate Unconjugated
Formulation In 100mM sodium phosphate buffer, 0.1% BSA, pH 7.0
Host Species Mouse
Immunogen a synthetic peptide (VTPRDKPMGPLLVATF) corresponding to amino acids 468-483 of human MMP-2
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽72kDa latent and the ∽66kDa active forms of the MMP-2.
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.