Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-2 Mouse anti-Bovine, Horse, Human, Mouse, Pig, Rat, Rabbit, Clone: 42-5D11, MilliporeSigma™

Catalog No. MAB3308MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3308MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3308MI Supplier MilliporeSigma Supplier No. MAB3308
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-MMP-2 Antibody, a.a. 468-483 hMMP2, clone 42-5D11 is validated for use in ELISA, immunohistochemistry (paraffin), western blotting for the detection of MMP-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 42-5D11
Conjugate Unconjugated
Formulation Liquid in 0.1Msodium phosphate buffer, pH 7.0 containing 2% protease free bovine Serum albumin.
Gene Accession No. NM_004530.2
Gene Symbols MMP2, TBE-1, CLG4A, CLG4, MMP-II, MONA, MMP-2, EC 3.4.24.24
Host Species Mouse
Immunogen Synthetic oligopeptide corresponding to amino acid residue 468-483 (VTPRDKPMGPLLVATF) of human matrix metalloproteinase 2 (human MMP-2, gelatinase A).
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Equine, Human, Mouse, Pig, Rabbit, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.