Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-2 Mouse anti-Human, Clone: A-Gel VC2, MilliporeSigma™

Catalog No. MAB13431MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB13431MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB13431MI Supplier MilliporeSigma Supplier No. MAB13431
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MMP-2 Antibody, pro and active form, clone A-Gel VC2 is an antibody against MMP-2 for use in ELISA, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-2
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone A-Gel VC2
Conjugate Unconjugated
Formulation 10mM PBS, pH 7.4, with 0.2% BSA and 15mM sodium azide
Gene Accession No. NM_004530.2
Gene Symbols MMP2, TBE-1, CLG4A, CLG4, MMP-II, MONA, MMP-2, EC 3.4.24.24
Host Species Mouse
Immunogen Human APMA-activated native 72kDa Gelatinase A.
Quantity 500 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.