Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-2 Proform Mouse anti-Human, Mouse, Rat, Clone: CA-4001, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB13405
Description
Anti-MMP-2 Proform Antibody, N-terminus, clone CA-4001 detects level of MMP-2 Proform and has been published and validated for use in western blot, immunohistochemistry (paraffin).
Specifications
MMP-2 Proform | |
Monoclonal | |
Unconjugated | |
NM_004530.2 | |
Mouse | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Immunohistochemistry (Paraffin), Western Blot | |
CA-4001 | |
Purified from ascites fluid by Protein G chromatography. Liquid in 10mM PBS, pH 7.4, with 0.2% BSA and 15mM sodium azide | |
MMP2, TBE-1, CLG4A, CLG4, MMP-II, MONA, MMP-2, EC 3.4.24.24 | |
N-terminal peptide APSPIIKFPGDVAPKTDK of procollagenase IV, coupled to KLH.{Marqulies, et al 1992} | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction