Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-MMP-3 Clone: 55-2A4,

Catalog No. IM36100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
IM36100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IM36100UG Supplier MilliporeSigma™ Supplier No. IM36100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects MMP-3 Clone: 55-2A4 in Human, Porcine, Rat, Sheep samples, and it is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immuno Blotting

Matrix metalloproteinases (MMPs) are a family of enzymes that are responsible for the degradation of extracellular matrix components such as collagen, laminin and proteoglycans. In addition to sequence homology, all MMPs share the following characteristics: the catalytic mechanism is dependent upon a zinc ion at the active center, they cleave one or more extracellular matrix components, they are secreted as zymogens which are activated by removal of an approximately 10kDa segment from the N-terminus and they are inhibited by tissue inhibitor of metalloproteinases (TIMP). These enzymes are involved in normal physiological processes such as embryogenesis and tissue remodeling and may play an important role in arthritis, periodontitis, and metastasis. MMP-3 (stromelysin-1) is secreted as a 59/57kDa zymogen that is proteolytically processed to the 45 and 28kDa active forms. This enzyme degrades a wide spectrum of extracellular matrix proteins, including cartilage proteoglycans, fibronectin, type IV and IX collagen and laminin. MMP-3 is thought to play an important role in pathophysiological degradation processes associated with conditions such as rheumatoid arthritis and cancer cell invasion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-3
Applications Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 55-2A4
Conjugate Unconjugated
Formulation In 100mM sodium phosphate buffer, 0.1% BSA, pH 7.0.
Host Species Mouse
Immunogen human pro-MMP-3 from the conditioned media of rheumatoid synovial fibroblasts
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽57/59kDa latent and the ∽28/45kDa active forms of MMP-3. Will also recognize MMP-3/TIMP-1 complexes. Slightly cross-reacts (∽34%) with human MMP-10.
Target Species Human, Pig, Rat, Sheep
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.