Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-MMP-3 (Human), Clone: 4B7.3, Invitrogen™
Applications: ELISA, IHC, IP, WB
Supplier: Thermo Scientific OMA108030
Description
Host Species: Rabbit
Clone: 4B7.3
Cross Reactivity: Hu
Immunogen: Native human MMP-3.
Enzyme Immunoassay, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications
MMP-3 (Human) | |
4B7.3 | |
Unconjugated | |
Native human MMP-3. | |
100 μg | |
Human |
Monoclonal | |
1mg/mL | |
Rabbit | |
Affinity Purified | |
Primary |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction