Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-9 Mouse anti-Human, Clone: GE-213, MilliporeSigma™

Catalog No. MAB13415MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB13415MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB13415MI Supplier MilliporeSigma Supplier No. MAB13415
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MMP-9 Antibody, clone GE-213 is an antibody against MMP-9 for use in immunoprecipitation, western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-9
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone GE-213
Conjugate Unconjugated
Formulation Purified from ascites fluid by Protein G chromatography. Liquid in 10mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.
Gene Accession No. NM_004994.2
Gene Symbols MMP9, MMP-9, GELB, CLG4B, EC 3.4.24.35 [Contains: 67 kDa matrix metalloproteinase-9, 82 kDa matrix metalloproteinase-9].
Host Species Mouse
Immunogen Purified human gelatinase B.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.