Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-9 Rabbit anti-Human, Rat, Monkey, Primate, Horse, Canine, Pig, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB6001
Description
Anti-MMP-9 Antibody, N-terminus is an antibody against MMP-9 for use in western blotting.
Specifications
MMP-9 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Canine, Equine, Human, Monkey, Pig, Primate, Rat |
Western Blot | |
Unconjugated | |
MMP9; CLG4B; GELB; MMP-9 | |
KLH-conjugated synthetic peptide corresponding to the N-terminus of MMP-9. | |
100 μg | |
NP_004985.2 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction