Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-9 Rabbit anti-Guinea Pig, Human, Polyclonal, MilliporeSigma™

Catalog No. AB13458MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB13458MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB13458MI Supplier MilliporeSigma Supplier No. AB13458

Rabbit Polyclonal Antibody

Anti-MMP-9 Antibody detects level of MMP-9 and has been published and validated for use in western blot, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-9
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunogobulin by Protein A chromatography. Prepared in 10mM PBS, pH 7.4, with 0.2% BSA and 15mM sodium azide.
Gene Symbols MMP9; MMP-9; GELB; CLG4B
Host Species Rabbit
Immunogen Amino acids 626-644 of human MMP-9
Quantity 500 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_004994.2
Target Species Guinea Pig, Human
Content And Storage 2-8°C for up to 12 months.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.