Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

matrix metallopeptidase 10 (stromelysin 2), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017857 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-857 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-857 Supplier Abnova Corporation Supplier No. H00004319D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human MMP10 protein.

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The enzyme encoded by this gene degrades proteoglycans and fibronectin. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. [provided by RefSeq

Sequence: MMHLAFLVLLCLPVCSAYPLSGAAKEEDSNKDLAQQYLEKYYNLEKDVKQFRRKDSNLIVKKIQGMQKFLGLEVTGKLDTDTLEVMRKPRCGVPDVGHFSSFPGMPKWRKTHLTYRIVNYTPDLPRDAVDSAIEKALKVWEEVTPLTFSRLYEGEADIMISFAVKEHGDFYSFDGPGHSLAHAYPPGPGLYGDIHFDDDEKWTEDASGTNLFLVAAHELGHSLGLFHSANTEALMYPLYNSFTELAQFRLSQDDVNGIQSLYGPPPASTEEPLVPTKSVPSGSEMPAKCDPALSFDAISTLRGEYLFFKDRYFWRRSHWNPEPEFHLISAFWPSLPSYLDAAYEVNSRDTVFIFKGNEFWAIRGNEVQAGYPRGIHTLGFPPTIRKIDAAVSDKEKKKTYFFAADKYWRFDENSQSMEQGFPRLIADDFPGVEPKVDAVLQAFGFFYFFSGSSQFEFDPNARMVTHILKSNSWLHC

Specifications

Antigen matrix metallopeptidase 10 (stromelysin 2)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human MMP10 protein.
Formulation PBS with no preservative; pH 7.4
Gene MMP10
Gene Accession No. NM_002425
Gene Alias SL-2/STMY2
Gene Symbols MMP10
Host Species Rabbit
Immunogen MMP10 (NP_002416.1, 1 a.a. ∼ 476 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4319
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.