Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP17 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA513186
Description
This antibody is predicted to react with mouse based on sequence homology.
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. MMP17 is considered a member of the membrane-type MMP (MT-MMP) subfamily. However, this protein is unique among the MT-MMPs in that it is a GPI-anchored protein rather than a transmembrane protein. The protein activates MMP-2 by cleavage.
Specifications
MMP17 | |
Polyclonal | |
MMP17 | |
MMP17 | |
KLH conjugated synthetic peptide between 391-420 amino acids from the central region of human MMP17 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9ULZ9 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
4326 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction