Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mn-SOD Rabbit anti-Human, Rat, Mouse, Bovine, Polyclonal, MilliporeSigma™

Catalog No. 06984MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-984-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-984-MI Supplier MilliporeSigma Supplier No. 06984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Mn-SOD Antibody is an antibody against Mn-SOD for use in immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mn-SOD
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 70% storage buffer 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide and 30% glycerol.
Gene Symbols SOD2; Mn-SOD; MNSOD; MnSOD; IPO-B
Host Species Rabbit
Immunogen Recombinant human MnSOD (manganous superoxide dismutase).
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_000627
Target Species Bovine, Human, Mouse, Rat
Content And Storage 2-8°C for up to 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.