Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MNAT1, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89200389 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-200-389 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-200-389 Supplier Abnova Corporation Supplier No. H00004331K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human MNAT1 peptide using ARM Technology.

Cyclin-dependent kinases (CDKs), which play an essential role in cell cycle control of eukaryotic cells, are phosphorylated and thus activated by the CDK-activating kinase (CAK). CAK is a multisubunit protein that includes CDK7 (MIM 601955), cyclin H (CCNH; MIM 601953), and MAT1. MAT1 (for 'menage a trois-1') is involved in the assembly of the CAK complex.[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen MNAT1
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description menage a trois homolog 1, cyclin H assembly factor (Xenopus laevis)
Formulation In 1X PBS, pH 7.4
Gene MNAT1
Gene Accession No. MNAT1
Gene Alias MAT1, RNF66, TFB3
Gene Symbols MNAT1
Host Species Rabbit
Immunogen A synthetic peptide of human MNAT1
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4331
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.