Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MnSOD Mouse anti-Human, Rat, Rabbit, Clone: MnS-1, MilliporeSigma™

Catalog No. MAB4081MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4081MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4081MI Supplier MilliporeSigma Supplier No. MAB4081
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-MnSOD Antibody, clone MnS-1 is validated for use in western blot, immunocytochemistry, immunohistochemistry for the detection of MnSOD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MnSOD
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MnS-1
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS.
Gene Accession No. NM_000636.2; NM_001024465.1; NM_001024466.1
Gene Symbols SOD2, MNSOD, MnSOD, IPO-B, Mn-SOD, EC 1.15.1.1
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rabbit, Rat
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.