Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOB1, Mps One Binder kinase activator-like 2C (yeast), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016624 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-624 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-624 Supplier Abnova Corporation Supplier No. H00148932B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human MOBKL2C protein.

The protein encoded by this gene is similar to the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq

Sequence: MALCLKQVFAKDKTFRPRKRFEPGTQRFELYKKAQASLKSGLDLRSVVRLPPGENIDDWIAVHVVDFFNRINLIYGTMAERCSETSCPVMAGGPRYEYRWQDERQYRRPAKLSAPRYMALLMDWIEGLINDEEVFPTRVGVPFPKNFQQVCTKILTRLFRVFVHVYIHHFDSILSMGAEAHVNTCYKHFYYFIREFSLVDQRELEPLREMTERICH

Specifications

Antigen MOB1, Mps One Binder kinase activator-like 2C (yeast)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MOBKL2C protein.
Formulation PBS with no preservative; pH 7.4
Gene MOBKL2C
Gene Accession No. NM_201403.1
Gene Alias MGC26743/MOB3C
Gene Symbols MOBKL2C
Host Species Mouse
Immunogen MOBKL2C (NP_958805.1, 1 a.a. ∼ 216 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Kinases
Primary or Secondary Primary
Gene ID (Entrez) 148932
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.