Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mono/di/trimethyl-Histone H3 (Lys4) Rabbit anti-Human, Clone: AW304, MilliporeSigma™

Catalog No. 05791RMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-791-RMI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-791-RMI Supplier MilliporeSigma Supplier No. 05791R

Rabbit Monoclonal Antibody

Anti-mono/di/trimethyl-Histone H3 (Lys4) Antibody, clone AW304 is a rabbit monoclonal antibody for detection of mono/di/trimethyl-Histone H3 (Lys4) also known as histone H3.1, H3 histone family member I and has been validated in western blot, PIA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mono/di/trimethyl-Histone H3 (Lys4)
Applications Inhibition Assays, Western Blot
Classification Monoclonal
Clone AW304
Conjugate Unconjugated
Formulation Purified Rabbit Monoclonal IgG containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl, 40% Glycerol and 0.05% sodium azide.
Gene Accession No. NP_066298
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Histone H3 mono/di/tri-methylated at Lys4.
Purification Method Protein A
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) HIST1H3A, H3FA, HIST1H3B, H3FL, HIST1H3C, H3FC, HIST1H3D, H3FB, HIST1H3F, H3FI, HIST1H3G, H3FH, HIST1H3H, H3FK, HIST1H3I, H3FF, HIST1H3J, H3FJ
Target Species Human
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.