Learn More
Monocarboxylate Transporter 1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Monocarboxylate Transporter 1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Liquid in PBS containing 0.1% BSA . |
| Gene Symbols | SLC16A1; MCT; FLJ36745; MGC44475; MCT1 |
| Host Species | Rabbit |
| Immunogen | A 17 amino acid peptide sequence near the C-terminus of rat MCT1 (Gerhart et al. 1997; Knott et al. 1999; Takanaga et al. 1995; Carpenter et al. 1996; Koehler-Stec et al. 1998; Yoon et al. 1999; Halestrap & Price 1999). |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.