Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monocarboxylate Transporter 4 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB3314PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3314PMI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3314PMI Supplier MilliporeSigma Supplier No. AB3314P
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Monocarboxylate Transporter 4 using this Anti-Monocarboxylate Transporter 4 Antibody validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Monocarboxylate Transporter 4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA and 0.05% sodium azide.
Gene Symbols SLC16A3; MCT3; MGC138472; MGC138474; MCT4
Host Species Rabbit
Immunogen A 19 amino acid peptide sequence near the C-terminus of rat/human/mouse MCT4.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001042422.1; NM_001042423.1; NM_004207.2
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.