Learn More
Monocarboxylate Transporter 8 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Monocarboxylate Transporter 8 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinitypurified immunoglobulin presented as a liquid in PBS with 0.1% BSA. |
| Gene Symbols | SLC16A2; DXS128E; DXS128; MCT8; AHDS; XPCT; MCT7 |
| Host Species | Rabbit |
| Immunogen | A 19 amino acid peptide sequence within the cytoplasmic, C-terminus of mouse MCT8. The immunogen sequence is 94% conserved in rat and 89% in human MCT8 and shows no significant sequence homology with other MCT family members. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.