Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monocyte Chemotactic Protein-1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1834P
Description
Anti-Monocyte Chemotactic Protein-1 Antibody is an antibody against Monocyte Chemotactic Protein-1 for use in ELISA, western blot, NEUT.
Specifications
Monocyte Chemotactic Protein-1 | |
Polyclonal | |
Affinity purified immunoglobulin. Lyophilized from 0.5X PBS pH 7.4. Contains no preservative. Reconstitute with 50μL sterile distilled water. | |
Rabbit | |
Affinity Purified | |
Primary | |
Rat |
ELISA, Neutralization, Western Blot | |
Unconjugated | |
CCL2; SMC-CF; MCP1; MCAF; HSMCR30; GDCF-2; SCYA2; HC11; MGC9434; MCP-1 | |
Highly pure (≥98%) recombinant rat MCP-1 | |
50 μg | |
NP_113718 | |
Lyophilized: 2-8°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction