Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monocyte/Granulocyte Mouse anti-Pig, PE, Clone: 74-22-15A, BD

Catalog No. BDB561499 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561499 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561499 Supplier BD Biosciences Supplier No. 561499
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 74-22-15A (switch variant of 74-22-15) monoclonal antibody, an isotype class-switch variant of mAb 74-22-15, specifically binds to a 230-kDa protein expressed by most pig macrophages, peripheral blood monocytes and granulocytes, and few lymphocytes. mAb 74-22-15A does not crossreact with human or bovine cells. This clone was clustered as anti-SWC3a at the First International Swine CD workshop.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Monocyte/Granulocyte
Applications Flow Cytometry
Classification Monoclonal
Clone 74-22-15A
Concentration 0.2mg/mL
Conjugate PE
Description SWC3; CD172a
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen dd miniature swine thymocytes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Pig
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.