Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monomethyl-Histone H3 (Lys36) Rabbit anti-Vertebrates, Polyclonal, MilliporeSigma™

Catalog No. 07548MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-548-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-548-MI Supplier MilliporeSigma Supplier No. 07548
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-monomethyl-Histone H3 (Lys36) Antibody (Rabbit Polyclonal Antibody) validated in DB, immunofluoresence, western blot, Mplex to detect monomethyl-Histone H3 (Lys36) also known as H3K36me1, Histone H3 (monomethyl K36).
TRUSTED_SUSTAINABILITY

Specifications

Antigen monomethyl-Histone H3 (Lys36)
Applications Multiplex, Immunofluorescence, Immunoassay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.05% sodium azide and 30% glycerol
Gene Symbols H3F3A; MGC87783; H3.3A; MGC87782; H3F3; H3.3B; H3F3B
Host Species Rabbit
Immunogen Peptide containing the sequence GVmeKKP corresponding to monomethyl Lysine 36 of human histone H3 .
Purification Method Antiserum
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Target Species Vertebrates
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.