Learn More
Monomethyl-Histone H3 (Lys4) Rabbit anti-Human, Vertebrates, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | monomethyl-Histone H3 (Lys4) |
| Applications | ChIP sequencing (ChIP-seq), Dot Blot, Immunocytochemistry, Immunofluorescence, Inhibition Assays, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit IgG in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05 Sodium Azide, pH 7.4. |
| Gene Symbols | HIST3H3; H3FT; H3/t; H3t; MGC126888; H3/g; H3.4; H3T; MGC126886 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated, synthetic peptide containing the sequence ART[meK]..., in which meK corresponds to monomethyl-lysine at residue 4 of human histone H3. |
| Purification Method | Protein A purfied |
| Quantity | 200 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.