Learn More
Monomethyl-Histone H3 (Lys9) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | monomethyl-Histone H3 (Lys9) |
| Applications | Dot Blot, Immunocytochemistry, Inhibition Assays, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. |
| Gene Symbols | HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated branched peptide corresponding to human Histone H3 methylated at Lys9. |
| Purification Method | Affinity Purified |
| Quantity | 10 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.