Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monomethyl-Histone H4 (Lys20) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$144.24 - $615.02
Specifications
Antigen | monomethyl Histone H4 (Lys20) |
---|---|
Applications | ChIP Assay, Dot Blot, Inhibition Assays, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
07-157-0SMI
|
MilliporeSigma
071570S |
10 μg |
Each for $144.24
|
|
|||||
07-157-0MI
|
MilliporeSigma
071570 |
100 μg |
Each for $615.02
|
|
|||||
Description
Anti-monomethyl Histone H4 (Lys20) Antibody is a rabbit polyclonal antibody for detection of monomethyl Histone H4 (Lys20) also known as H4K20me1, Histone H4 (mono methyl K20), H4 histone family member A and has been validated in western blotting.Specifications
monomethyl Histone H4 (Lys20) | |
Polyclonal | |
Rabbit | |
Purified from serum by affinity chromatography. Supplied at 1mg/mL in 0.1M Tris-Glycine (pH7.4), 15mM NaCl, 0.05% NaN3. | |
H4/A; H4/B; H4/C; H4/D; H4/E; H4/G ; H4/H; H4/I ; H4/J ; H4/K ; H4/M; H4/N; H4/O; H4/a ; H4F2; H4FA ; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; HIST2H4 | |
Primary | |
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles |
ChIP Assay, Dot Blot, Inhibition Assays, Western Blot | |
Unconjugated | |
Human | |
NM_003538.3 | |
KLH-conjugated, linear synthetic peptide corresponding to amino acids 15-24 (AKRHR[me1K]VLRD-C) of human Histone H4., linear synthetic peptide corresponding to amino acids 15-24 (AKRHR[me1K]VLRD-C) of human Histone H4. |
|
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title