Learn More
Monoubiquitin H2A Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | monoubiquitin H2A (Lys119) |
| Applications | ChIP Assay, Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 90mM Tris-glycine pH8, 50mM Nacl, and 10% glycerol with 0.05% sodium azide |
| Gene Symbols | HTA2 |
| Host Species | Rabbit |
| Immunogen | Branch peptide corresponding to human monoubiquitin H2A (Lys119). |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.