Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB4140
Description
Anti-MOP3 Antibody detects level of MOP3 and has been published and validated for use in western blot, immunocytochemistry, immunohistochemistry.
Specifications
MOP3 | |
Polyclonal | |
Affinity purified immunoglobulin precipitated in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
SBNO1; FLJ10701; MOP3; mop-3; Drosophila; FLJ10833; MOP-3; MOP3.; Sno; FLJ16176 | |
Synthetic peptide corresponding to amino acids 257-272 of human MOP3. (SVKVEDKDFPSTCSKK). | |
100 μg | |
NM_018183.2 | |
-20°C for up to 6 months, Avoid Freeze/Thaw Cycles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction