Learn More
v-mos Moloney murine sarcoma viral oncogene homolog, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MOS protein.
Supplier: Abnova Corporation H00004342B01
Description
MOS is a serine/threonine kinase that activates the MAP kinase cascade through direct phosphorylation of the MAP kinase activator MEK (MAP2K1; MIM 176872) (Prasad et al., 2008 [PubMed 18246541]).[supplied by OMIM
Sequence: MPSPLALRPYLRSEFSPSVDARPCSSPSELPAKLLLGATLPRAPRLPRRLAWCSIDWEQVCLLQRLGAGGFGSVYKATYRGVPVAIKQVNKCTKNRLASRRSFWAELNVARLRHDNIVRVVAASTRTPAGSNSLGTIIMEFGGNVTLHQVIYGAAGHPEGDAGEPHCRTGGQLSLGKCLKYSLDVVNGLLFLHSQSIVHLDLKPANILISEQDVCKISDFGCSEKLEDLLCFQTPSYPLGGTYTHRAPELLKGEGVTPKADIYSFAITLWQMTTKQAPYSGERQHILYAVVAYDLRPSLSAAVFEDSLPGQRLGDVIQRCWRPSAAQRPSARLLLVDLTSLKAELGSpecifications
v-mos Moloney murine sarcoma viral oncogene homolog | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human MOS protein. | |
MOS | |
MGC119962/MGC119963/MSV | |
Mouse | |
50 μL | |
Cell Cycle | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_005372.1 | |
MOS | |
MOS (NP_005363.1, 1 a.a. ∼ 346 a.a) full-length human protein. | |
RUO | |
Yes | |
4342 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.