Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Fodrin Antibody (3D7), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192689AF647
Description
Alpha Fodrin Monoclonal specifically detects Alpha Fodrin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Alpha Fodrin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Q13813 | |
| SPTAN1 | |
| Recombinant construct (expressed in and purified from E. coli) containing the 19th and 20th spectrin repeats and C-terminal EF hand domains of human alpha Fodrin (amino acids 2086-2447) [UniProt# Q13813]. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 3D7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2 | |
| Mouse | |
| 240 kDa | |
| 0.1 mL | |
| Neuroscience | |
| 6709 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction