Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-APLP-1, Clone: plpc1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP150309
Description
Myelin PLP Monoclonal antibody specifically detects Myelin PLP in Human, Bovine samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Myelin PLP | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml, Immunohistochemistry-Frozen 1:10-1:500, Immunofluorescence | |
PLP1 | |
Synthetic peptide GRGTKF corresponding to C terminal region of myelin proteolipid protein. | |
0.05 mg | |
Immune System Diseases, Immunology, Neuroscience, Stem Cell Markers | |
5354 | |
Human, Bovine | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence | |
plpc1 | |
Unconjugated | |
HLD1, lipophilin, major myelin proteolipid protein, MMPL, myelin proteolipid protein, PLP, PLP/DM20, PMD, proteolipid protein 1, spastic paraplegia 2, uncomplicated, SPG2 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Recognizes myelin proteolipid protein (PLP) in many mammalian species. This Recognizes DM20. PLP encodes the major protein components of compact CNS myelin and mutations in the PLP gene can lead to severe dysmyelinating disease | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction