Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Beclin 1/ATG6, Biotin, Clone: 4H10, Novus Biologicals™

Supplier: Novus Biologicals NBP100085B
Specifications
Beclin 1 | |
Monoclonal | |
Biotin | |
Q14457 | |
BECN1 | |
Full length human Beclin 1 protein [Swiss-Prot# Q14457] | |
Protein G purified | |
RUO | |
Primary | |
Human. 99% sequence identity with monkey, 98% identity with mouse and bovine, 97% identity with porcine, rat and horse, 91% identity with chicken. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
4H10 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30 | |
Mouse | |
52 kDa | |
0.1 mL | |
Apoptosis, Autophagy, Cancer, Hypoxia, Immunology, Innate Immunity, mTOR Pathway, Tumor Suppressors | |
8678 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction