Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase-3 Antibody (31A1067), PE - (Pro and Active), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056708PE
Description
Caspase-3 Monoclonal specifically detects Caspase-3 in Human, Mouse, Rat, Porcine, Chicken, Chinese Hamster, Mammal samples. It is validated for Flow Cytometry, Knockout Validated, Knockdown Validated.Specifications
Caspase-3 | |
Monoclonal | |
PE | |
P42574 | |
CASP3 | |
This Caspase-3 Antibody (31A1067) - (Pro and Active) was developed against full-length recombinant human caspase-3 protein. | |
Protein G purified | |
RUO | |
Primary | |
The antibody detects both pro Caspase-3 (∼32 kDa) and the large subunit of the active/cleaved form (∼14-21 kDa) of Caspase-3. | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunoassay, KnockDown | |
31A1067 | |
Flow Cytometry, Knockout Validated, Knockdown Validated | |
Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
Mouse | |
31.7 kDa | |
0.1 mL | |
Apoptosis, Cancer, Caspases, Phospho Specific | |
836 | |
Human, Mouse, Rat, Pig, Chicken, Chinese Hamster, Mammalia | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction