Learn More
Description
Host Species: Rat
Clone: APA5
Isotype: IgG2a, κ
Cross Reactivity Species: Ms
Immunogen: Mouse PDGF Receptor α chain
Flow Cytometry
Specifications
Specifications
| Antigen | Mouse CD140A |
| Classification | Monoclonal |
| Clone | APA5 |
| Concentration | 0.2mg/mL |
| Conjugate | PE |
| Host Species | Rat |
| Immunogen | Mouse PDGF Receptor α chain |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.