Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD282 Rat anti-Mouse, Alexa Fluor 647, Clone: 6C2, BD

Catalog No. BDB562625 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562625 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562625 Supplier BD Biosciences Supplier No. 562625
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 6C2 monoclonal antibody specifically binds to the mouse Toll like receptor 2 (TLR2) that is also known as CD282. CD282 is a type 1 transmembrane glycoprotein and a TLR family member. It is expressed on the surface of monocytes, macrophages, dentritic cells, granulocytes, endothelial and epithelial cells. This pattern recognition receptor is important for sensing and transducing signals in response to microbial cell wall components including lipoproteins, peptidoglycans and lipoteichoic acid. CD282 signals through intracellular MYD88 and TRAF6 leading to NF-kappa-B activation and the production of cytokines and inflammatory mediators by responding cells. TLR2 plays a major role in mediating inflammatory and innate immune responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD282
Applications Flow Cytometry
Classification Monoclonal
Clone 6C2
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description TLR2; Tlr2; Toll-like receptor 2; Ly105
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse TLR2 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.