Learn More
CD36 Mouse anti-Mouse, PE, Clone: CRF D-2712, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 562702
Description
The CRF D-2712 monoclonal antibody specifically binds to mouse and rat CD36, an 88-kDa glycoprotein member of the class B scavenger receptor family. CD36 is a multifunctional receptor that has a variety of ligands and is expressed on many cell types. Consequently, it is known by several names, including macrophage scavenger receptor, platelet glycoprotein IV, and fatty acid translocase. Cells that express CD36 include platelets, megakaryocytes, monocytes/macrophages, dendritic cells, erythroid precursors, microvascular endothelia, skeletal, cardiac, and smooth muscle cells, adipocytes, and epithelia of retina, breast, and intestine. This multi-ligand receptor functions in the clearance of apoptotic cells and abnormal erythrocytes (in malaria and sickle-cell anemia), the transport of long-chain fatty acids and oxidized low-density lipoproteins, the regulation of angiogenesis by mediating the effects of thrombospondin-1, and the mediation of coronary vasoconstriction. Crosslinking of cell-surface CD36 with plate-bound CRF D-2712 antibody promotes internalization of photoreceptor outer segment fragments by cultured retinal pigment epithelium.

Specifications
CD36 | |
Monoclonal | |
0.2mg/mL | |
Platelet glycoprotein 4; PAS-4; GPIIIB; Fatty acid translocase; Scavenger R | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgA κ |
Flow Cytometry | |
CRF D-2712 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant mouse CD36 | |
0.1 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.