Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD38 Rat anti-Mouse, Brilliant Violet 421, Clone: 90/CD38, BD

Catalog No. BDB562768 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562768 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562768 Supplier BD Biosciences Supplier No. 562768
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 90 monoclonal antibody specifically binds to CD38, a 42 kDa transmembrane glycoprotein on immature and mature, resting and activated, B lymphocytes. In contrast to humans, CD38 expression is down-regulated on mouse germinal center B cells and plasma cells. CD38 is also expressed on a subpopulation of thymic and peripheral T cells, NK cells, and splenic macrophages. Furthermore, CD38 has been detected on bone marrow-derived hematopoietic stem cells. The CD38 molecule is reported to exhibit both cyclase and hydrolase activities and plays a role in lymphocyte activation. CD31, both human and mouse, is reported to be a ligand for human CD38.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD38
Applications Flow Cytometry
Classification Monoclonal
Clone 90/CD38
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description ADP-ribosyl cyclase 1; Cyclic ADP-ribose hydrolase 1; I-19; NIM-R5
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Bone Marrow Pre-B cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.