Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD38 Antibody (1G7F4) - Azide Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP225265

 View more versions of this product

Catalog No. NBP225265


Only null left
Add to Cart

Description

Description

CD38 Monoclonal specifically detects CD38 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

CD38
Monoclonal
0.5mg/mL
Western Blot 3 - 5 ug/mL, Simple Western, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence reported by customer review, Immunohistochemistry-Paraffin 5 ug/mL, Immunohistochemistry-Frozen
CD38
Amino acids 71-197 of mouse CD38 were used as the immunogen for the antibody.
0.1 mg
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers
952
Human, Mouse
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1G7F4
Unconjugated
ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10
Mouse
Protein G purified
RUO
Primary
The amino acid sequence used as immunogen for the antibody is 100% homologous in Mouse, 91% in Rat and 64% in Human.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2b λ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only