Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse CD38, Fluorescein, Polyclonal, R&D Systems™

Catalog No. FAB4947F
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4947F 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4947F Supplier R&D Systems Supplier No. FAB4947F
This item is not returnable. View return policy

Host Species: Sheep
Species Reactivity:Mouse
Immunogen:Mouse myeloma cell line NS0-derived recombinant mouse CD38, Leu45-Thr304, Accession No. EDL37596

Applications:

Flow Cytometry

Specifications

Antigen CD38
Applications Flow Cytometry
Classification Polyclonal
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. EDL37596
Gene Alias ADP-ribosyl Cyclase, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose Hydrolase, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10
Gene Symbols CD38
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD38, Leu45-Thr304, Accession # EDL37596
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 952
Test Specificity Detects mouse CD38 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant human CD38 is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.