Learn More
Description
Host Species: Armenian Hamster
Clone: HM48-1
Isotype: IgG1, λ3
Cross Reactivity Species: Ms
Immunogen: Mouse T lymphoma MBL-2
Flow Cytometry
Specifications
Specifications
| Antigen | Mouse CD48 |
| Classification | Monoclonal |
| Clone | HM48-1 |
| Concentration | 0.2mg/mL |
| Conjugate | APC |
| Host Species | Hamster |
| Immunogen | Mouse T lymphoma MBL-2 |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.