Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD52 Antibody (HI186), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500311PE
Description
CD52 Monoclonal specifically detects CD52 in Human samples. It is validated for Flow Cytometry.Specifications
CD52 | |
Monoclonal | |
PE | |
P31358 | |
CD52 | |
Human tonisl (NM_001803.2). | |
0.1 mL | |
Cancer, Immunology | |
1043 | |
Human | |
IgG2b |
Flow Cytometry | |
HI186 | |
Flow Cytometry | |
Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
The antibody HI186 reacts with CD52 (CAMPATH-1), a 21-28 kDa glycoprotein containing a large N-linked carbohydrate moiety; mature CD52 molecule is actually much smaller (approx. 8-9 kDa). CD52 is expressed at high levels on lymphocytes, monocytes/macrophages and in male reproductive tract. HLDA VI; WS Code BP 523 HLDA VI; WS Code T 6T-057 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction