Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (RPA-T8), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP225195AF405
Description
CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Flow (Cell Surface), Product Image.Specifications
CD8 | |
Monoclonal | |
Alexa Fluor 405 | |
NP_741969 | |
CD8A | |
CD8 (CD8a; alpha chain) | |
Protein G purified | |
RUO | |
Primary | |
This product reacts with Human. | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
RPA-T8 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Flow (Cell Surface) | |
CD8, CD8a molecule, LEU2, p32 | |
Mouse | |
26 kDa | |
0.1 mL | |
Adaptive Immunity, Cytokine Research, Immunology, Phospho Specific, Signal Transduction, Stem Cell Markers | |
925 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction