Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPA Antibody (5A7-2E11), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212892
Description
CENPA Monoclonal specifically detects CENPA in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| CENPA | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence | |
| Centromere autoantigen A, centromere protein A (17kD), centromere protein ACENP-Acentromere protein A, 17kDa, histone H3-like centromeric protein A | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 1058 | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 5A7-2E11 | |
| Unconjugated | |
| NP_001035891.1 | |
| CENPA | |
| Synthetic peptide corresponding to a portion of human CENP-A | |
| 0.1 mg | |
| Primary | |
| Detects approx 18kDa. Recognizes Human CENP-A. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction