Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clathrin Heavy Chain 1/CHC17 Antibody (X22), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300613PE
Description
Clathrin Heavy Chain 1/CHC17 Monoclonal specifically detects Clathrin Heavy Chain 1/CHC17 in Human, Mouse, Rat, Bovine, Hamster, Primate samples. It is validated for Flow Cytometry.Specifications
Clathrin Heavy Chain 1/CHC17 | |
Monoclonal | |
PE | |
Q00610 | |
CLTC | |
Purified human brain clathrin heavy chain. | |
0.1 mL | |
Membrane Trafficking and Chaperones, Membrane Vesicle Markers | |
1213 | |
Human, Mouse, Rat, Bovine, Hamster, Primate | |
Purified |
Flow Cytometry | |
X22 | |
Flow Cytometry | |
CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Detects clathrin heavy chain. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction