Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL12/SDF-1 Antibody (MM0211-9N26), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212221APC
Description
CXCL12/SDF-1 Monoclonal specifically detects CXCL12/SDF-1 in Human, Rat, Sheep samples. It is validated for Flow Cytometry.Specifications
CXCL12/SDF-1 | |
Monoclonal | |
APC | |
Q6ICW0 | |
CXCL12 | |
Human recombinant CXCL12/SDF-1 | |
0.1 mL | |
Growth and Development, Neuronal Cell Markers | |
6387 | |
Human, Rat, Sheep | |
Purified |
Flow Cytometry | |
MM0211-9N26 | |
Flow Cytometry | |
chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects human CXCL12 (both 1-alpha and 1 beta) | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction