Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 13 Antibody (2D7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197797
Description
Cytokeratin 13 Monoclonal specifically detects Cytokeratin 13 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoblotting.Specifications
Cytokeratin 13 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2 with 0.09% Sodium Azide | |
KRT13 | |
Clone 2D7 is a mouse monoclonal IgG2b antibody derived by fusion of SP2/0 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a cytokeratin preparation extracted from human esophagus. | |
1 mL | |
Primary | |
Clone 2D7 reacts exclusively with cytokeratin 13 which is present in non-cornified squamous epithelia, except cornea, and transitional epithelial regions, with the exception of basal cell layers of some stratified epithelia. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoblot | |
2D7 | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoblotting | |
CK13, CK-13, cytokeratin 13, Cytokeratin-13, K13cytokeratin-13, keratin 13, keratin, type I cytoskeletal 13, keratin-13, MGC161462, MGC3781 | |
Mouse | |
Tissue culture supernatant | |
RUO | |
3860 | |
Human, Mouse, Rat, Rabbit | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction