Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digoxin Antibody (25P1C9), Novus Biologicals™
SDP

Catalog No. NBP178617 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178617 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178617 Supplier Novus Biologicals Supplier No. NBP178617

Mouse Monoclonal Antibody

Digoxin Monoclonal specifically detects Digoxin in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Radioimmunoassay, Chemiluminescence Immunoassay.

Specifications

Antigen Digoxin
Applications Western Blot, ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Chemiluminescence Immunoassay (CIA)
Classification Monoclonal
Clone 25P1C9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, Chemiluminescence Immunoassay
Gene Alias Lanoxin
Host Species Mouse
Immunogen Digoxin-BSA
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Chemical.
Target Species Other
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.