Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Mouse DNA-PKcs, Clone: 18-2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001203
Description
DNA-PKcs Monoclonal specifically detects DNA-PKcs in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
DNA-PKcs | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 100, Immunofluorescence 1:10 - 1:500 | |
DNA-dependent protein kinase, DNA-dependent protein kinase catalytic subunit-like, DNA-PK catalytic subunit, DNA-PKcs, DNPK1, EC 2.7.11.1, HYRC, HYRC1, p460 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
18-2 | |
Unconjugated | |
P78527 | |
PRKDC | |
Human DNA-dependent Protein Kinase purified from HeLa cells | |
500 μL | |
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining, Protein Kinase | |
5591 | |
Store at 4C. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction