Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUX4 Antibody (P4H2), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP149552UV
Description
DUX4 Monoclonal specifically detects DUX4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
DUX4 | |
Monoclonal | |
DyLight 350 | |
Q9UBX2 | |
DUX4 | |
Synthetic peptide corresponding to the C-terminus of human DUX4. [Swiss-Prot: Q9UBX2] | |
Protein G purified | |
RUO | |
Primary | |
This antibody is specific for DUX4 and does not recognize DUX4c. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
P4H2 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
double homeobox 4, double homeobox protein 10, double homeobox protein 4, double homeobox protein 4/10, double homeobox protein DUX10, DUX10 | |
Mouse | |
52 kDa | |
0.1 mL | |
Neuroscience, Transcription Factors and Regulators | |
100288687 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction