Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FALDH Antibody (OTI2A7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP201017
Description
FALDH Monoclonal specifically detects FALDH in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FALDH | |
Monoclonal | |
0.62 mg/mL | |
Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
ALDH3A2 | |
Full length human recombinant protein of human ALDH3A2(NP_001026976) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Rat | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI2A7 | |
Unconjugated | |
Aldehyde dehydrogenase 10, aldehyde dehydrogenase 3 family, member A2, Aldehyde dehydrogenase family 3 member A2, ALDH10FLJ20851, EC 1.2.1, EC 1.2.1.3, FALDHDKFZp686E23276, fatty aldehyde dehydrogenase, Microsomal aldehyde dehydrogenase, SLS | |
Mouse | |
57.5 kDa | |
0.1 mL | |
Stem Cell Markers | |
224 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction