Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-9 Antibody (FG9-77) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB600623
Description
FGF-9 Monoclonal specifically detects FGF-9 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot, Microarray.Specifications
| FGF-9 | |
| Monoclonal | |
| 2.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Dot Blot 5 ug/ml, Microarray | |
| FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3 | |
| Mouse | |
| 23 kDa | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Cycle and Replication | |
| 2254 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot | |
| FG9-77 | |
| Unconjugated | |
| P54130 | |
| FGF9 | |
| Recombinant mouse FGF-9 | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Monoclonal Anti-FGF9 reacts specifically with recombinant FGF9 (fibroblast growth factor 9). It does not cross-react with human FGF Acidic and FGF Basic. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction